Tisk…

1,2,3-Trifluorobenzene

MOLEM42703898EA 0 CZK
MOLEM42703898
1,2,3-Trifluorobenzene
1,2,3-Trifluorobenzene
Vzorec: C₆H₃F₃
MW: 132,09 g/mol
Bod varu: 95 °C (1013 hPa)
Hustota: 1,28 g/cm³ (20 °C)
MDL Number: MFCD00012013
CAS číslo: 1489-53-8
UN: 2387
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR