3-Iodobenzoic acid
MOLEM77238188EA
0
CZK
MOLEM77238188
3-Iodobenzoic acid
3-Iodobenzoic acid
Vzorec:
C₇H₅IO₂ MW: 248,02 g/mol Bod varu: 337 °C (1013 hPa) Bod tání: 186…188 °C Hustota: 2,171 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002496 CAS číslo: 618-51-9 EINECS: 210-555-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...