Tisk…

ALFAA15385

Dodavatel: Thermo Scientific
A15385.22EA 0 CZK
ALFAA15385.22 ALFAA15385.14
ALFAA15385
Purity: 98%
CAS No.: 122-95-2
Molecular Formula: C10H14O2
Formula Weight: 166.22
Melting Point: 69-71°
Boiling Point: 245-248°
MDL No.: MFCD00015146
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR