L-Fenylalanin, Supelco®
Dodavatel: Merck
Synonyms:
L-α-Fenylalanin
91331-25GEA
2360
CZK
91331-25G
L-Fenylalanin, Supelco®
L-Fenylalanin
Vzorec:
C₉H₁₁NO₂ MW: 165,19 g/mol Bod varu: 313 °C (1013 hPa) Bod tání: 173 °C Hustota: 0,754 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064227 CAS číslo: 63-91-2 EINECS: 200-568-1 Merck Index: 14,07271 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...