Tisk…

4-Bromo-3-nitrobenzoic acid, Sigma-Aldrich®

Dodavatel: Merck

SIAL724734-5GEA 601 CZK
SIAL724734-5G
4-Bromo-3-nitrobenzoic acid, Sigma-Aldrich®
4-Bromo-3-nitrobenzoic acid
Vzorec: C₇H₄BrNO₄
MW: 246,02 g/mol
MDL Number: MFCD00272137
CAS číslo: 6319-40-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR