Tisk…

4,4'-Bis(dimethylamino)thiobenzophenone, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL515388-5GEA 2880 CZK
SIAL515388-5G
4,4'-Bis(dimethylamino)thiobenzophenone, Sigma-Aldrich®
4,4'-Bis(dimethylamino)thiobenzophenone
Vzorec: C₁₇H₂₀N₂S
MW: 284,43 g/mol
MDL Number: MFCD00040477
CAS číslo: 1226-46-6
EINECS: 214-962-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR