(±)-2-Bromoisovaleric acid, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
SIAL251003-25GEA
2920
CZK
SIAL251003-25G
(±)-2-Bromoisovaleric acid, Sigma-Aldrich®
(±)-2-Bromoisovaleric acid
Vzorec:
C₅H₉BrO₂ MW: 181,03 g/mol |
MDL Number:
MFCD00004210 CAS číslo: 565-74-2 EINECS: 209-291-0 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...