Tisk…

DL-α-(Methylaminomethyl)benzyl alcohol, Sigma-Aldrich®

Dodavatel: Merck

SIAL209848-10GEA 0 CZK
SIAL209848-10G
DL-α-(Methylaminomethyl)benzyl alcohol, Sigma-Aldrich®
DL-α-(Methylaminomethyl)benzyl alcohol
Vzorec: C₉H₁₃NO
MW: 151,21 g/mol
MDL Number: MFCD00004506
CAS číslo: 6589-55-5
EINECS: 229-525-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR