Tisk…

(±)-1-(1-Naphthyl)ethylamine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL294926-5GEA 0 CZK
SIAL294926-5G
(±)-1-(1-Naphthyl)ethylamine, Sigma-Aldrich®
(±)-1-(1-Naphthyl)ethylamine
Vzorec: C₁₂H₁₃N
MW: 171,24 g/mol
MDL Number: MFCD00004014
CAS číslo: 42882-31-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR