Tisk…

α-Ionone, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIALI12409-100GEA 2210 CZK
SIALI12409-100G
α-Ionone, Sigma-Aldrich®
α-Ionone
Vzorec: C₁₃H₂₀O
MW: 192,3 g/mol
MDL Number: MFCD00001565
CAS číslo: 127-41-3
EINECS: 204-841-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR