1-Methyl-2-phenylindole, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL404888-10GEA
1050
CZK
SIAL404888-10G
1-Methyl-2-phenylindole, Sigma-Aldrich®
1-Methyl-2-phenylindole
Vzorec:
C₁₅H₁₃N MW: 207,27 g/mol |
MDL Number:
MFCD00022892 CAS číslo: 3558-24-5 EINECS: 222-618-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...