Tisk…

4-Nitro-3-(trifluoromethyl)benzonitrile, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL590185-5GEA 6010 CZK
SIAL590185-5G
4-Nitro-3-(trifluoromethyl)benzonitrile, Sigma-Aldrich®
4-Nitro-3-(trifluoromethyl)benzonitrile
Vzorec: C₈H₃F₃N₂O₂
MW: 216,12 g/mol
MDL Number: MFCD05664270
CAS číslo: 320-36-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR