Tisk…

1-Benzyloxy-3-iodobenzene, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL532193-10GEA 5450 CZK
SIAL532193-10G
1-Benzyloxy-3-iodobenzene, Sigma-Aldrich®
1-Benzyloxy-3-iodobenzene
Vzorec: C₁₃H₁₁IO
MW: 310,13 g/mol
MDL Number: MFCD01318100
CAS číslo: 107623-21-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR