Tisk…
Serological pipettes, disposable
Dodavatel: Bürkle
BURK7556-0050EA 132 CZK
BURK7556-0050
Serological pipettes, disposable
PS, sterile.
  • Crystal clear
  • For aqueous solutions
  • Individually packed
These disposable pipettes are ideal for training purposes, in industrial labs, during field research and for applications where pipettes made of glass cannot be used, e.g. in flourine and silicate chemistry.