Grade 8 ruled filter papers, Whatman™
Supplier: Whatman products (Cytiva)
Certificates
About this item
These white cotton linter cellulose filters are ideal for routine investigations of foreign substances in a variety of sample types.
- Resistant to aqueous and non aqueous solutions
Printed green lines for optical assessment at 5 mm intervals.
Specifications
- Catalog No:
- WHAT10347008
- WHAT10347004
- WHAT10347033
- WHAT10347009
- Weight:
- 65 g/m²
- 65 g/m²
- 65 g/m²
- 65 g/m²
- Particle retention:
- 20 - 30 µm
- 20 - 30 µm
- 20 - 30 µm
- 20 - 30 µm
Specifications
Product Information
- Catalog #Supplier #GradeSizePkAvailabilityPrice
- Specifications:
More
Specifications
Environmentally PreferableWeight65 g/m²Particle retention20 - 30 µm - Specifications:
More
Specifications
Environmentally PreferableWeight65 g/m²Particle retention20 - 30 µm - Specifications:
More
Specifications
Environmentally PreferableWeight65 g/m²Particle retention20 - 30 µm - Specifications:
More
Specifications
Environmentally PreferableWeight65 g/m²Particle retention20 - 30 µm