Cyclopentadecanone
Dodavatel: EHRENSTORFER GMBH
EHERC11832000EA
1750
CZK
EHERC11832000
Cyclopentadecanone
Cyclopentadecanone
Vzorec:
C₁₅H₂₈O MW: 224,39 g/mol Bod varu: 338 °C (1013 hPa) Bod tání: 63 °C Hustota: 0,89 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001266 CAS číslo: 502-72-7 EINECS: 207-951-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...