Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Czech Republic
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0,00 Kč
Order Entry
Home
Saccharin sodium
Saccharin sodium
Saccharin sodium
Saccharin sodium
EDQMS0050000
:
EDQM
:
Saccharin sodium
EDQMS0050000
:
EDQM
:
: S0050000
: EDQMS0050000
6 990,00 Kč
1 * 50 mg
This product has been blocked by your organization. Please contact your purchasing department for more information
Pack type:
Glass bottle for solids
Pk:
50 mg
Seq:
2370
:
:
206.18 g/mol
:
204-886-1
:
128-44-9
:
C₇H₅NNaO₃S
:
MFCD00013092