Retinol all trans
MOLEM21418841EA
0
CZK
MOLEM21418841
Retinol all trans
Retinol all trans
Vzorec:
C₂₀H₃₀O MW: 286,46 g/mol |
MDL Number:
MFCD00001552 CAS číslo: 68-26-8 EINECS: 200-683-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...