2,6-Dichloroisonicotinic acid
MOLE63506294-1GEA
0
CZK
MOLE63506294-1G
2,6-Dichloroisonicotinic acid
2,6-Dichloroisonicotinic acid
Vzorec:
C₆H₃Cl₂NO₂ MW: 192 g/mol |
MDL Number:
MFCD00234147 CAS číslo: 5398-44-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...