Tisk…

Silikagel 15% in water, colloidal dispersion

Dodavatel: Thermo Scientific
Varování

36669.A7EA 3950 CZK
36669.A7 36669.A1
Silikagel 15% in water, colloidal dispersion
Silikagel

High-temperature binders, anti-slip coatings for paper, petrochemical catalyst support and binder, polishing agent, abrasion resistant coatings

Vzorec: SiO₂
MW: 60,08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS číslo: 7631-86-9
EINECS: 231-545-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR