Tisk…
Gaskets for flat flanges, BOLA
Dodavatel: Bohlender
BOLA
BOHLB282-04EA 3220 CZK
BOHLB282-04 BOHLB282-02 BOHLB321-01
Gaskets for flat flanges, BOLA
Těsnění Těsnění
PTFE, white.
  • Temperature resistance: from −60 to +230 °C
  • Universal chemical resistance
  • Vacuum: suitable
Gasket with silicone inlet (thickness 0,5 mm). The product is only exposed to PTFE.