Kyselina mravenčí 98-100% ACS, Reagent Grade
Dodavatel: SCHARLAB S.L.
|
Nebezpečí
|
SCARAC10851000EA
0
CZK
CHESAC10852500
SCARAC10852500
SCARAC10851000
SCARAC1085025P
Kyselina mravenčí 98-100% ACS, Reagent Grade
Kyselina mravenčí
Vzorec:
HCO₂H MW: 46,03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003297 CAS číslo: 64-18-6 EINECS: 200-579-1 UN: 1779 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...