Cleanroom gloves, latex, sterile, BioClean Maxima™
Supplier: Nitritex
Certificates
About this item
These cleanroom gloves with beaded cuffs can be used in medical as well as in laboratory environments. Suitable for double-donning.
- Sterile, powder-free, textured finish
- Protein level less than 50 µg/g
- Latex provides exceptional flexibility and comfort
- Anatomically shaped for enhanced wearer comfort
- Extended length provides full arm protection
- Each size carries a unique barcode on the carton (ITF-14), which allows automatic data entry into stock control systems
Suitable for use in Class 10, ISO 4, M2.5 and EU GMP Grade A cleanrooms.
Certifications: Manufactured in a facility holding ISO 9001:2015 and (EU)2016/425 Module D certifications.
Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Regulation (EU)2016/425.
Processed in an NEBB certified ISO Class 4 environment.
Complies with the requirements of European Glove Standards EN 420:2003 +A1:2009, EN 374-1:2016, EN 374-5:2016 and EN 421:2010.
Physical properties comply with European Medical Glove Standard EN 455-2:2015.
Sterilized in accordance with ISO 11137-1:2006.
Suitable for contact with all foodstuffs. Regulation (EC)1935/2004 and (EC) 2023/2006.
Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Regulation (EU)2016/425.
Processed in an NEBB certified ISO Class 4 environment.
Complies with the requirements of European Glove Standards EN 420:2003 +A1:2009, EN 374-1:2016, EN 374-5:2016 and EN 421:2010.
Physical properties comply with European Medical Glove Standard EN 455-2:2015.
Sterilized in accordance with ISO 11137-1:2006.
Suitable for contact with all foodstuffs. Regulation (EC)1935/2004 and (EC) 2023/2006.
Packaging: One cuffed pair per inner plastic wallet; one wallet per EasyTear™ PE pouch; 10 pairs per sealed outer PE bag; 10 outer bags per carton liner; one carton liner per carton (100 pairs).
Specifications
- Catalog No:
- 112-3847
- 112-3845
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Disposable
- Disposable
- Powdered / powder-free:
- Powder-free
- Powder-free
- Cleanroom class:
- 10/ISO 4; EU GMP (A)
- 10/ISO 4; EU GMP (A)
- Cuff style:
- Beaded
- Beaded
- Cuff thickness:
- 0,12 mm
- 0,12 mm
- Finger thickness:
- 0,22 mm
- 0,22 mm
- Palm thickness:
- 0,20 mm
- 0,20 mm
- EN norm:
- EN 455, EN 374
- EN 455, EN 374
Specifications
Product Information
- Catalog #Supplier #Glove sizeColourLengthPackedPkAvailabilityPrice
- Specifications:
More
Specifications
Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableDisposablePowdered / powder-freePowder-freeCleanroom class10/ISO 4; EU GMP (A)Cuff styleBeadedCuff thickness0,12 mmFinger thickness0,22 mmPalm thickness0,20 mmEN normEN 455, EN 374 - Specifications:
More
Specifications
Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableDisposablePowdered / powder-freePowder-freeCleanroom class10/ISO 4; EU GMP (A)Cuff styleBeadedCuff thickness0,12 mmFinger thickness0,22 mmPalm thickness0,20 mmEN normEN 455, EN 374