Tisk…
Grounding mat
Dodavatel: SCAT Europe
SCTV108099EA 4820 CZK
SCTV108099
Grounding mat
Rohože Rohože podlahové, permanentní
Anti-static mat, conductive. With ground cable (press stud connector).
  • Several mats can be connected to each other
Thickness: 3 mm