Tisk…

(1R,2R)-(+)-1,2-Diphenylethylenediamine

Dodavatel: ABCR GMBH

ABCRAB168484.0005EA 0 CZK
ABCRAB168484.0005
(1R,2R)-(+)-1,2-Diphenylethylenediamine
(1R,2R)-(+)-1,2-Diphenylethylenediamine
Vzorec: C₁₄H₁₆N₂
MW: 212,29 g/mol
MDL Number: MFCD00082769
CAS číslo: 35132-20-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR