Order Entry
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
 :  Merck
undefined
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier:  Merck
   Sustainable Options Available
Certificates
  • M-Clarity quality level:
    MQ100
  • Environmentally Preferable:
    Sustainable Packaging
Product Information
  • Catalog #
    Supplier #
    pH value
    pH buffer type
    Pack type
    Pk
    Availability
    Price
  • Buffer solution (citric acid/sodium hydroxide/hydrogen chloride), traceable to SRM from NIST and PTB pH 2.00 (20 °C) Certipur®
    Buffer solution (citric acid/sodium hydroxide/hydrogen chloride), traceable to SRM from NIST and PTB pH 2.00 (20 °C) Certipur®
    1.09433.1000
    1.09433.1000
    2,00
    Citrate
    Plastic bottle
    1 L
    Log in for availability
    1 * 1 L
    1 * 1 L - 1 800,00 Kč
    1 800,00 Kč
    1 * 1 L
       1 800,00 Kč
  • Buffer solution (citric acid/sodium hydroxide/hydrogen chloride), traceable to SRM from NIST and PTB pH 2.00 (20 °C) Certipur®
    Buffer solution (citric acid/sodium hydroxide/hydrogen chloride), traceable to SRM from NIST and PTB pH 2.00 (20 °C) Certipur®
    1.09433.4000
    1.09433.4000
    2,00
    Citrate
    Titripac®
    4 L
    Log in for availability
    1 * 4 L
    1 * 4 L - 5 950,00 Kč
    5 950,00 Kč
    1 * 4 L
       5 950,00 Kč
    Specifications:
      Sustainable Packaging
      Sustainable Packaging
  • Buffer solution (citric acid/sodium hydroxide/hydrogen chloride), traceable to SRM from NIST and PTB pH 3.00 (20 °C) Certipur®
    Buffer solution (citric acid/sodium hydroxide/hydrogen chloride), traceable to SRM from NIST and PTB pH 3.00 (20 °C) Certipur®
    1.09434.1000
    1.09434.1000
    3,00
    Citrate
    Plastic bottle
    1 L
    Log in for availability
    1 * 1 L
    1 * 1 L - 1 800,00 Kč
    1 800,00 Kč
    1 * 1 L
       1 800,00 Kč
  • Buffer solution (citric acid/sodium hydroxide/hydrogen chloride), traceable to SRM from NIST and PTB pH 4.00 (20 °C) Certipur®
    Buffer solution (citric acid/sodium hydroxide/hydrogen chloride), traceable to SRM from NIST and PTB pH 4.00 (20 °C) Certipur®
    1.09435.1000
    1.09435.1000
    4,00
    Citrate
    Plastic bottle
    1 L
    Log in for availability
    1 * 1 L
    1 * 1 L - 1 800,00 Kč
    1 800,00 Kč
    1 * 1 L
       1 800,00 Kč
  • Buffer solution (citric acid/sodium hydroxide/hydrogen chloride), traceable to SRM from NIST and PTB pH 4.00 (20 °C) Certipur®
    Buffer solution (citric acid/sodium hydroxide/hydrogen chloride), traceable to SRM from NIST and PTB pH 4.00 (20 °C) Certipur®
    1.09435.4000
    1.09435.4000
    4,00
    Citrate
    Titripac®
    4 L
    Log in for availability
    1 * 4 L
    1 * 4 L - 5 950,00 Kč
    5 950,00 Kč
    1 * 4 L
       5 950,00 Kč
    Specifications:
      Sustainable Packaging
      Sustainable Packaging
  • Buffer solution (acetic acid/sodium acetate), traceable to SRM from NIST and PTB pH 4.66 (20 °C) Certipur®
    Buffer solution (acetic acid/sodium acetate), traceable to SRM from NIST and PTB pH 4.66 (20 °C) Certipur®
    1.07827.1000
    1.07827.1000
    4,66
    Acetate
    Plastic bottle
    1 L
    Log in for availability
    1 * 1 L
    1 * 1 L - 2 190,00 Kč
    2 190,00 Kč
    1 * 1 L
       2 190,00 Kč
  • Buffer solution (citric acid/sodium hydroxide), traceable to SRM from NIST and PTB pH 5.00 (20 °C) Certipur®
    Buffer solution (citric acid/sodium hydroxide), traceable to SRM from NIST and PTB pH 5.00 (20 °C) Certipur®
    1.09436.1000
    1.09436.1000
    5,00
    Citrate
    Plastic bottle
    1 L
    Log in for availability
    1 * 1 L
    1 * 1 L - 1 800,00 Kč
    1 800,00 Kč
    1 * 1 L
       1 800,00 Kč
  • Buffer solution (citric acid/sodium hydroxide), traceable to SRM from NIST and PTB pH 6.00 (20 °C) Certipur®
    Buffer solution (citric acid/sodium hydroxide), traceable to SRM from NIST and PTB pH 6.00 (20 °C) Certipur®
    1.09437.1000
    1.09437.1000
    6,00
    Citrate
    Plastic bottle
    1 L
    Log in for availability
    1 * 1 L
    1 * 1 L - 1 800,00 Kč
    1 800,00 Kč
    1 * 1 L
       1 800,00 Kč
  • Buffer solution (citric acid/sodium hydroxide), traceable to SRM from NIST and PTB pH 6.00 (20 °C) Certipur®
    Buffer solution (citric acid/sodium hydroxide), traceable to SRM from NIST and PTB pH 6.00 (20 °C) Certipur®
    1.09437.4000
    1.09437.4000
    6,00
    Citrate
    Titripac®
    4 L
    Log in for availability
    1 * 4 L
    1 * 4 L - 5 950,00 Kč
    5 950,00 Kč
    1 * 4 L
       5 950,00 Kč
    Specifications:
      Sustainable Packaging
      Sustainable Packaging
  • Buffer solution (di-sodium hydrogen phosphate/potassium hydrogen phosphate) traceable to SRM from NIST and PTB pH 6.88 (20°C) CertiPUR®
    Buffer solution (di-sodium hydrogen phosphate/potassium hydrogen phosphate) traceable to SRM from NIST and PTB pH 6.88 (20°C) CertiPUR®
    1.07294.1000
    1.07294.1000
    6,88
    Phosphate
    Plastic bottle
    1 L
    Log in for availability
    1 * 1 L
    1 * 1 L - 2 190,00 Kč
    2 190,00 Kč
    1 * 1 L
       2 190,00 Kč
  • Buffer solution (disodium hydrogen phosphate/potassium dihydrogen phosphate) traceable to SRM from NIST and PTB pH 7.00 (20 °C) Certipur®
    Buffer solution (disodium hydrogen phosphate/potassium dihydrogen phosphate) traceable to SRM from NIST and PTB pH 7.00 (20 °C) Certipur®
    1.09439.1000
    1.09439.1000
    7,00
    Phosphate
    Plastic bottle
    1 L
    Log in for availability
    1 * 1 L
    1 * 1 L - 1 800,00 Kč
    1 800,00 Kč
    1 * 1 L
       1 800,00 Kč
  • Buffer solution (disodium hydrogen phosphate/potassium dihydrogen phosphate) traceable to SRM from NIST and PTB pH 7.00 (20 °C) Certipur®
    Buffer solution (disodium hydrogen phosphate/potassium dihydrogen phosphate) traceable to SRM from NIST and PTB pH 7.00 (20 °C) Certipur®
    1.09439.4000
    1.09439.4000
    7,00
    Phosphate
    Titripac®
    4 L
    Log in for availability
    1 * 4 L
    1 * 4 L - 5 950,00 Kč
    5 950,00 Kč
    1 * 4 L
       5 950,00 Kč
    Specifications:
      Sustainable Packaging
      Sustainable Packaging
  • Buffer solution (boric acid/sodium hydroxide/hydrogen chloride), traceable to SRM from NIST and PTB pH 8.00 (20 °C) Certipur®
    Buffer solution (boric acid/sodium hydroxide/hydrogen chloride), traceable to SRM from NIST and PTB pH 8.00 (20 °C) Certipur®
    1.09460.1000
    1.09460.1000
    8,00
    Borate
    Plastic bottle
    1 L
    Log in for availability
    1 * 1 L
    1 * 1 L - 1 800,00 Kč
    1 800,00 Kč
    1 * 1 L
       1 800,00 Kč
  • Buffer solution (boric acid/potassium chloride/sodium hydroxide), traceable to SRM from NIST and PTB pH 9.00 (20 °C) Certipur®
    Buffer solution (boric acid/potassium chloride/sodium hydroxide), traceable to SRM from NIST and PTB pH 9.00 (20 °C) Certipur®
    1.09461.1000
    1.09461.1000
    9,00
    Borate
    Plastic bottle
    1 L
    Log in for availability
    1 * 1 L
    1 * 1 L - 1 800,00 Kč
    1 800,00 Kč
    1 * 1 L
       1 800,00 Kč
  • Buffer solution (boric acid/potassium chloride/sodium hydroxide), traceable to SRM from NIST and PTB pH 9.00 (20 °C) Certipur®
    Buffer solution (boric acid/potassium chloride/sodium hydroxide), traceable to SRM from NIST and PTB pH 9.00 (20 °C) Certipur®
    1.09461.4000
    1.09461.4000
    9,00
    Borate
    Titripac®
    4 L
    Log in for availability
    1 * 4 L
    1 * 4 L - 5 950,00 Kč
    5 950,00 Kč
    1 * 4 L
       5 950,00 Kč
    Specifications:
      Sustainable Packaging
      Sustainable Packaging
  • Buffer solution (di-sodium tetraborate), traceable to SRM from NIST and PTB pH 9.22 (20°C) CertiPUR®
    Buffer solution (di-sodium tetraborate), traceable to SRM from NIST and PTB pH 9.22 (20°C) CertiPUR®
    1.01645.1000
    1.01645.1000
    9,22
    Borate
    Plastic bottle
    1 L
    Log in for availability
    1 * 1 L
    1 * 1 L - 2 190,00 Kč
    2 190,00 Kč
    1 * 1 L
       2 190,00 Kč
  • Buffer solution (boric acid/potassium chloride/sodium hydroxide), traceable to SRM from NIST and PTB pH 10.00 (20 °C) Certipur®
    Buffer solution (boric acid/potassium chloride/sodium hydroxide), traceable to SRM from NIST and PTB pH 10.00 (20 °C) Certipur®
    1.09438.1000
    1.09438.1000
    10,00
    Borate
    Plastic bottle
    1 L
    Log in for availability
    1 * 1 L
    1 * 1 L - 1 800,00 Kč
    1 800,00 Kč
    1 * 1 L
       1 800,00 Kč
  • Buffer solution (boric acid/potassium chloride/sodium hydroxide), traceable to SRM from NIST and PTB pH 10.00 (20 °C) Certipur®
    Buffer solution (boric acid/potassium chloride/sodium hydroxide), traceable to SRM from NIST and PTB pH 10.00 (20 °C) Certipur®
    1.09438.4000
    1.09438.4000
    10,00
    Borate
    Titripac®
    4 L
    Log in for availability
    1 * 4 L
    1 * 4 L - 5 950,00 Kč
    5 950,00 Kč
    1 * 4 L
       5 950,00 Kč
    Specifications:
      Sustainable Packaging
      Sustainable Packaging
  • Buffer solution (boric acid/potassium chloride/sodium hydroxide), traceable to SRM from NIST and PTB pH 11.00 (20 °C) Certipur®
    Buffer solution (boric acid/potassium chloride/sodium hydroxide), traceable to SRM from NIST and PTB pH 11.00 (20 °C) Certipur®
    1.09462.1000
    1.09462.1000
    11,00
    Borate
    Plastic bottle
    1 L
    Log in for availability
    1 * 1 L
    1 * 1 L - 1 800,00 Kč
    1 800,00 Kč
    1 * 1 L
       1 800,00 Kč
  • Buffer solution (disodium hydrogen phosphate/sodium hydroxide) traceable to SRM from NIST und PTB pH 12.00 (25 °C) Certipur®
    Buffer solution (disodium hydrogen phosphate/sodium hydroxide) traceable to SRM from NIST und PTB pH 12.00 (25 °C) Certipur®
    1.99022.4000
    1.99022.4000
    12,00
    Phosphate
    Titripac®
    4 L
    Log in for availability
    1 * 4 L
    1 * 4 L - 5 950,00 Kč
    5 950,00 Kč
    1 * 4 L
       5 950,00 Kč