Order Entry
COD cell test for non Merck photometers, Spectroquant®, Supelco®
COD cell test for non Merck photometers, Spectroquant®, Supelco®
Supplier:  Merck
Certificates
undefined
COD cell test for non Merck photometers, Spectroquant®, Supelco®
Supplier:  Merck
Certificates
Specifications
  • M-Clarity quality level:
    MQ100
Specifications
Related Information
Product Information
  • Catalog #
    Supplier #
    Range
    Chemical method
    Pk
    Availability
    Price
  • COD cell test
    COD cell test
    1.18751.0001
    1.18751.0001
    0 - 150 mg/l
    Chromosulphuric oxidation
    25 Tests
    Log in for availability
    1 * 25 Tests
    1 * 25 Tests - 4 250,00 Kč
    4 250,00 Kč
    1 * 25 Tests
       4 250,00 Kč
     : 1.18751.0001
     : MERC1.18751.0001
  • COD cell test
    COD cell test
    1.18750.0001
    1.18750.0001
    0 - 40 mg/l
    Chromosulphuric oxidation
    25 Tests
    1 * 25 Tests
    1 * 25 Tests - 4 250,00 Kč
    4 250,00 Kč
    1 * 25 Tests
       4 250,00 Kč
     : 1.18750.0001
     : MERC1.18750.0001
  • COD cell test
    COD cell test
    1.18753.0001
    1.18753.0001
    0 - 15000 mg/l
    Chromosulphuric oxidation
    25 Tests
    1 * 25 Tests
    1 * 25 Tests - 5 160,00 Kč
    5 160,00 Kč
    1 * 25 Tests
       5 160,00 Kč
     : 1.18753.0001
     : MERC1.18753.0001
  • COD cell test
    COD cell test
    1.18752.0001
    1.18752.0001
    0 - 1500 mg/l
    Chromosulphuric oxidation
    25 Tests
    1 * 25 Tests
    1 * 25 Tests - 4 250,00 Kč
    4 250,00 Kč
    1 * 25 Tests
       4 250,00 Kč
     : 1.18752.0001
     : MERC1.18752.0001