Tisk…

Fenthion (O,O-Dimethyl-O-(4-methylthion-m-tolyl) phosphorothioate)

Dodavatel: EHRENSTORFER GMBH

EHERC13580000EA 1750 CZK
EHERC13580000
Fenthion (O,O-Dimethyl-O-(4-methylthion-m-tolyl) phosphorothioate)
Fenthion (O,O-dimethyl-O-(4-methylthion-m-tolyl)phosphorothioate)
Vzorec: C₁₀H₁₅O₃PS₂
MW: 278,33 g/mol
CAS číslo: 55-38-9
EINECS: 200-231-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR