Carbofuran
Dodavatel: EHRENSTORFER GMBH
EHERC11010000EA
2550
CZK
EHERC11010000
Carbofuran
Carbofuran
Vzorec:
C₁₂H₁₅NO₃ MW: 221,26 g/mol |
CAS číslo:
1563-66-2 EINECS: 216-353-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...