Tisk…
Ladies coats, cotton, short sleeves
Dodavatel: Uvex
UVEX8151.101EA 990 CZK
UVEX8151.101 UVEX8151.112 UVEX8151.111 UVEX8151.103 UVEX8151.102 UVEX8151.113 UVEX8151.110 UVEX8151.109 UVEX8151.108 UVEX8151.105 UVEX8151.104 UVEX8151.107 UVEX8151.106
Ladies coats, cotton, short sleeves
Pracovní oděvy
These coats with short sleeves are made of 100% cotton.
  • Revers collar
  • Concealed stainless steel push buttons
  • Two side pockets, one breast pocket
  • Slightly fitted waist
  • Comply to Öko-Tex Standard 100
Weight: 210 g/m²
Length: 105 cm