Tisk…
ESD safety shoes, lace-up, uvex office 9541
Dodavatel: Uvex
uvex office
UVEX9541.4/46EA 3440 CZK
UVEX9541.4/46 UVEX9541.4/47 UVEX9541.4/44 UVEX9541.4/45 UVEX9541.4/42 UVEX9541.4/43 UVEX9541.4/40 UVEX9541.4/41 UVEX9541.4/39 UVEX9541.4/38
ESD safety shoes, lace-up, uvex office 9541
Obuv
Elegant ESD office shoe in brogue style made from supple, full-grain leather upper and an antistatic nitrile rubber sole (cut and abrasion resistant, heat-resistant for brief exposure up to +300 °C; oil and petrol-resistant, highly acid and alkali-resistant)
  • Breathable lining
  • Ultra comfortable insole
  • Protective steel toecap (200 J)
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Colour: Black
Certifikace: EN 61340 (ESD); EN ISO 20345:2011; S1, SRA