Chcete se opět přihlásit?
Cleaning concentrates for laboratory, elma lab clean
Dodavatel: Elma
Laboratory cleaning agents designed to meet requirements for instrumentation and glassware in analysis laboratories. All Elma Lab Clean cleaning agents are formulated for use with ultrasonic intensive cleaning devices.
These products are also suitable for laboratory rinsing machines, as they are highly effective without foaming.
OBJEDNÁVKA
Rozbalit vše / Zavřít vše
Popis Objem Balené Bal. VWR katalogové číslo Množství
elam lab clean acidic concentrate, S10, for cleaning labware and instruments 1 l Box 1 l ELMA8000095
elam lab clean acidic concentrate, S10, for cleaning labware and instruments 10 l Box 10 l ELMA8000097
elam lab clean acidic concentrate, S10, for cleaning labware and instruments 2,5 l Box 2,5 l ELMA8000096
elam lab clean acidic concentrate, S10, for cleaning labware and instruments 25 l Box 25 l ELMA8000098
elam lab clean alkaline cleaning concentrate, A10, for ultrasonic baths and labware washers 1 l Box 1 l ELMA8000180
elam lab clean alkaline cleaning concentrate, A10, for ultrasonic baths and labware washers 10 l Box 10 l ELMA8000182
elam lab clean alkaline cleaning concentrate, A10, for ultrasonic baths and labware washers 2,5 l Box 2,5 l ELMA8000181
elam lab clean alkaline cleaning concentrate, A10, for ultrasonic baths and labware washers 25 l Box 25 l ELMA8000183
elam lab clean alkaline concentrate, A25, for cleaning in biological and medical labs, ultrasonic baths and labware washers 2,5 l Box 2,5 l ELMA8000086
elam lab clean alkaline concentrate, A25, for cleaning in biological and medical labs, ultrasonic baths and labware washers 25 l Box 25 l ELMA8000088
elam lab clean alkaline concentrate, A25, for cleaning in biological and medical labs, ultrasonic baths and labware washers 10 l Box 10 l ELMA8000087
elam lab clean alkaline concentrate, A25, for cleaning in biological and medical labs, ultrasonic baths and labware washers 1 l Box 1 l ELMA8000085
elma lab clean acidic concentrate, S20, for cleaning laboratory instruments 10 l Box 10 l ELMA8000192
elma lab clean acidic concentrate, S20, for cleaning laboratory instruments 2,5 l Box 2,5 l ELMA8000191
elma lab clean acidic concentrate, S20, for cleaning laboratory instruments 1 l Box 1 l ELMA8000190
elma lab clean acidic concentrate, S20, for cleaning laboratory instruments 25 l Box 25 l ELMA8000193
elma lab clean alkaline concentrate, A20sf, surfactant free, for cleaning ultrasonic baths, pipettes and labware washers 25 l Box 25 l ELMA8000058
elma lab clean alkaline concentrate, A20sf, surfactant free, for cleaning ultrasonic baths, pipettes and labware washers 1 l Box 1 l ELMA8000055
elma lab clean alkaline concentrate, A20sf, surfactant free, for cleaning ultrasonic baths, pipettes and labware washers 10 l Box 10 l ELMA8000057
elma lab clean alkaline concentrate, A20sf, surfactant free, for cleaning ultrasonic baths, pipettes and labware washers 2,5 l Box 2,5 l ELMA8000056
elma lab clean neutral concentrate, N10, for ultrasonic baths and labware washers 2,5 l Box 2,5 l ELMA8000076
elma lab clean neutral concentrate, N10, for ultrasonic baths and labware washers 10 l Box 10 l ELMA8000077
elma lab clean neutral concentrate, N10, for ultrasonic baths and labware washers 25 l Box 25 l ELMA8000078
elma lab clean neutral concentrate, N10, for ultrasonic baths and labware washers 1 l Box 1 l ELMA8000075
Generovat čárové kódy Formát popisku čárového kódu : 
Vygenerovat PDF katalog
PŘÍSLUŠENSTVÍ
Rozbalit vše / Zavřít vše
Accessories for cleaning concentrates
Popis Bal. VWR katalogové číslo Množství
Dosing pump, 30 ml, for 5 l and 10 l cans 1 ELMA8000003929
Tap for 25 l cans 1 ELMA8000003928
Tap for 5 l and 10 l cans 1 ELMA8000003927
Generovat čárové kódy Formát popisku čárového kódu : 
Vygenerovat PDF katalog
Enter Details
Select the article for which you are looking for Certificate
Doplňte Lot číslo. Toto pole je povinné. Jestli číslo neznáte, napíšte "x".
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Nebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Katalogové číslo Availability Jednotka Vaše cena Price Per Množství