Tisk…
Buffer, PBS solution (phosphate buffered saline), Ultra Pure Grade
Dodavatel: VWR Life Science
K812-20LEA 21410 CZK
K812-20L K812-500ML
Buffer, PBS solution (phosphate buffered saline), Ultra Pure Grade
Pufry Pufry pro elektroforézu Pufry promývací
Dulbecco's formulation without calcium and magnesium.
  • Free from DNase, RNase or protease
137,7 mM sodium chloride, 2,7 mM potassium chloride, 9,55 mM phosphate buffer
pH @ 25 °C      7,1 to 7,7