Tisk…
Tris-glycine-SDS (TGS) buffer solution, 10X liquid concentrate, Ultra Pure Grade
Dodavatel: VWR Life Science
0783-1LEA 1680 CZK
0783-1L 0783-4L 0783-5L
Tris-glycine-SDS (TGS) buffer solution, 10X liquid concentrate, Ultra Pure Grade
Pufry Pufry pro elektroforézu Pufry do dělícího gelu
Liquid concentrate used for protein electrophoresis to maintain pH and temperature of the environment as well as provide ions that carry electrical current.