Shelving starter and add-on units, MetroMax Q®
About this item
These Super Adjustable 2™ Starter and Add-On units save time and money by eliminating adjacent posts and replacing them with 'S' hooks. This system offers a convenient way to create long runs of shelving, shelving at right angles and combinations of runs and right angles.
The load capacity evenly distributed for shelf lengths up to 1219 mm is 363 kg per shelf and for shelf lengths 1524 mm and longer is 272 kg per shelf.
Chrome plated starter units includes four 1880 mm post and five 457 mm deep Super Adjustable 2™ shelves. Add-on units includes two 1880 mm post and five 457 mm deep Super Adjustable 2™ shelves and ten 'S' hooks (2 per shelf).
Product Information
- Catalog #Supplier #SekvencePopisŠ×H×VHmotnostBal.AvailabilityPrice
- METO5AQ337G35AQ337G301MetroMax Q® shelving add-on unit914×457×1880 mm25 kg11 * 1 KS25 770,00 KčSupplier Article Number: 5AQ337G3Local Article Number: METO5AQ337G3
Specifications
Environmentally Preferable - METO5AQ357G35AQ357G302MetroMax Q® shelving add-on unit1219×457×1880 mm32 kg11 * 1 KS29 760,00 KčSupplier Article Number: 5AQ357G3Local Article Number: METO5AQ357G3
Specifications
Environmentally Preferable - METO5AQ457G35AQ457G303MetroMax Q® shelving add-on unit1219×530×1880 mm36 kg11 * 1 KS32 520,00 KčSupplier Article Number: 5AQ457G3Local Article Number: METO5AQ457G3
Specifications
Environmentally Preferable - METO5AQ467G35AQ467G304MetroMax Q® shelving add-on unit1524×530×1880 mm44 kg11 * 1 KS36 660,00 KčSupplier Article Number: 5AQ467G3Local Article Number: METO5AQ467G3
Specifications
Environmentally Preferable - METO5AQ557G35AQ557G305MetroMax Q® shelving add-on unit1219×610×1880 mm39 kg11 * 1 KS35 090,00 KčSupplier Article Number: 5AQ557G3Local Article Number: METO5AQ557G3
Specifications
Environmentally Preferable - METO5AQ567G35AQ567G306MetroMax Q® shelving add-on unit1524×610×1880 mm48 kg11 * 1 KS40 360,00 KčSupplier Article Number: 5AQ567G3Local Article Number: METO5AQ567G3
Specifications
Environmentally Preferable - 132-05305Q337G307MetroMax Q® shelving starter unit914×457×1880 mm30 kg11 * 1 KS27 790,00 KčSupplier Article Number: 5Q337G3Local Article Number: METO5Q337G3
Specifications
Environmentally Preferable - METO5Q347G35Q347G308MetroMax Q® shelving starter unit1066×457×1880 mm33 kg11 * 1 KS31 780,00 KčSupplier Article Number: 5Q347G3Local Article Number: METO5Q347G3
Specifications
Environmentally Preferable - 132-05315Q357G309MetroMax Q® shelving starter unit1219×457×1880 mm36 kg11 * 1 KS31 780,00 KčSupplier Article Number: 5Q357G3Local Article Number: METO5Q357G3
Specifications
Environmentally Preferable - 132-05325Q367G310MetroMax Q® shelving starter unit1524×457×1880 mm43 kg11 * 1 KS36 200,00 KčSupplier Article Number: 5Q367G3Local Article Number: METO5Q367G3
Specifications
Environmentally Preferable - METO5Q437G35Q437G311MetroMax Q® shelving starter unit914×530×1880 mm32 kg11 * 1 KS31 070,00 KčSupplier Article Number: 5Q437G3Local Article Number: METO5Q437G3
Specifications
Environmentally Preferable - METO5Q457G35Q457G312MetroMax Q® shelving starter unit1219×530×1880 mm41 kg11 * 1 KS34 550,00 KčSupplier Article Number: 5Q457G3Local Article Number: METO5Q457G3
Specifications
Environmentally Preferable - METO5Q467G35Q467G313MetroMax Q® shelving starter unit1524×530×1880 mm48 kg11 * 1 KS38 680,00 KčSupplier Article Number: 5Q467G3Local Article Number: METO5Q467G3
Specifications
Environmentally Preferable - 132-05335Q537G314MetroMax Q® shelving starter unit914×610×1880 mm35 kg11 * 1 KS31 920,00 KčSupplier Article Number: 5Q537G3Local Article Number: METO5Q537G3
Specifications
Environmentally Preferable - 132-05345Q557G315MetroMax Q® shelving starter unit1219×610×1880 mm44 kg11 * 1 KS37 110,00 KčSupplier Article Number: 5Q557G3Local Article Number: METO5Q557G3
Specifications
Environmentally Preferable - 132-05355Q567G316MetroMax Q® shelving starter unit1524×610×1880 mm53 kg11 * 1 KS42 380,00 KčSupplier Article Number: 5Q567G3Local Article Number: METO5Q567G3
Specifications
Environmentally Preferable - METO5Q577G35Q577G317MetroMax Q® shelving starter unit1825×610×1880 mm62 kg11 * 1 KS48 650,00 KčSupplier Article Number: 5Q577G3Local Article Number: METO5Q577G3