Tisk…
Shirt, long sleeves, Match 185G
Dodavatel: Fristads Kansas Group
Fristads®
KWIN100579940XSEA 1720 CZK
KWIN100579940XS KWIN100579940S KWIN1005795392XL KWIN100579940XL KWIN1005795393XL KWIN1005799403XL KWIN1005799402XL KWIN100579940L KWIN100579940M KWIN100579539XL KWIN100579539S KWIN100579539L KWIN100579539M KWIN100579539XS
Shirt, long sleeves, Match 185G
Pracovní oděvy
Poplin shirt for men, manufactured from 60% cotton and 40% polyester.
  • Concealed snap fastener opening at neck
  • Three chest pockets, two with velcro fastener, one with zip fastener
  • Sleeve pocket
  • Side slits, lengthened back
Weight: 185 g/m²