Tisk…
Fall protection kit for roof work, Titan™
Dodavatel: Miller by Honeywell
139-0233EA 4970 CZK
DALL1011895
Fall protection kit for roof work, Titan™
Safety kit for roof work. Ideal for all work on flat and sloping roofs and for vertical ascent and descent. The kit contains:
  • Titan™ two point safety harness with safety loop at the back and front (harness loops also at the front)
  • Titan™ automatic guided safety harness
  • Anchorage line, 10 m; lanyard (karabiner hooks), 0,3 m long
Certifikace: EN 361, EN 353-2