Tisk…
Men's shirt, short sleeves, Match 200G
Dodavatel: Fristads Kansas Group
Match
113-07207EA 0 CZK
113-07207 113-07208 113-07209 113-07221 113-07222 113-07223 113-07202 113-07224 113-07225 113-07203 113-07204 113-07226 113-07205 113-07227 113-07228 113-07206 113-07220 113-07218 113-07219 113-07210 113-07211 113-07212 113-07213 113-07214 113-07216 113-07217
Men's shirt, short sleeves, Match 200G
Pracovní oděvy
Basic shirt with short sleeves, made of 60% cotton and 40% polyester.
  • Button-down collar
  • Two chest pockets with flaps
  • Snap fasteners
Weight: 200 g/m²