Order Entry
Pump sets for concentrated acids/alkalis
Pump sets for concentrated acids/alkalis
 :  BURKLE
undefined
Pump sets for concentrated acids/alkalis
 :  BURKLE
  •  
    Material
    Flow rate
    Immersion depth
    Tube Ø
    Pk
  • Drum pump set with 230 V motor
    Drum pump set with 230 V motor
    BURK5762-0200
    5762-0200
    PVDF
    80 l/min
    1000 mm
    41 mm
    1
    1 * 1 KS
    1 * 1 KS - 116 390,00 Kč
    116 390,00 Kč
    1 * 1 KS
       116 390,00 Kč
     : 5762-0200
     : BURK5762-0200
    Specifications
    Environmentally Preferable
  • Drum pump set with 230 V motor
    Drum pump set with 230 V motor
    BURK5762-0201
    5762-0201
    PVDF
    80 l/min
    1200 mm
    41 mm
    1
    1 * 1 KS
    1 * 1 KS - 122 220,00 Kč
    122 220,00 Kč
    1 * 1 KS
       122 220,00 Kč
     : 5762-0201
     : BURK5762-0201