forensicGEM® DNA extraction kits have been developed to advance capabilities in forensic profiling by offering higher throughput levels, in manual or automated workflows, as well as a reduction in sample processing time and consumable consumption.
forensicGEM® Blood kit is capable of extracting DNA from fresh liquid blood (fingerpricks), bloodstains on a range of surfaces, EDTA, citrate, and heparin stabilised blood. The method is quick and done in a closed tube with a simple heat cycle and short centrifugation step that works in conjunction with the buffer formulation to remove inhibitors common in human blood samples, without loss of yield.
forensicGEM® Blood kit is capable of extracting DNA from fresh liquid blood (fingerpricks), bloodstains on a range of surfaces, EDTA, citrate, and heparin stabilised blood. The method is quick and done in a closed tube with a simple heat cycle and short centrifugation step that works in conjunction with the buffer formulation to remove inhibitors common in human blood samples, without loss of yield.