Tisk…
Antipain dihydrochloride
Dodavatel: G-Biosciences
786-045EA 6710 CZK
786-045
Antipain dihydrochloride
Proteiny
Antipain hydrochloride inhibits Ca2+-dependent endopeptidases, including papain, trypsin-like serine proteases, some cysteine proteases and to a lesser extent plasmin. It has a higher specificity for trypsin and papain compared to leupeptin.