n-Tridecyl-β-D-maltoside
MOLE54413003-1GEA
0
CZK
MOLE54413003-1G
n-Tridecyl-β-D-maltoside
n-Tridecyl-β-D-maltoside
Vzorec:
C₂₅H₄₈O₁₁ MW: 524,64 g/mol |
MDL Number:
MFCD08062386 CAS číslo: 93911-12-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...