Tisk…
Mitomycin C (from Streptomyces caespitosus), Sigma-Aldrich®
Dodavatel: Merck
Nebezpečí
M-Clarity™ quality level = MQ200
SIALM4287-5X2MGEA 30120 CZK
SIALM4287-5X2MG SIALM4287-2MG
Mitomycin C (from Streptomyces caespitosus), Sigma-Aldrich®
Mitomycin C
Vzorec: C₁₅H₁₈N₄O₅
MW: 334,33 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00078109
CAS číslo: 50-07-7
EINECS: 200-008-6
UN: 2811
ADR: 6.1,II