Order Entry
Permanent marker set, dual-tip, Sharpie®
  HEATHS15094
 :  HEATHROW
undefined
Permanent marker set, dual-tip, Sharpie®
  HEATHS15094
 :  HEATHROW
1 660,00 Kč
  1 * 8 KS
  • Colour:
    Assorted
  • Description:
    Permanent marker set, dual-tip, water-resistant ink
  • Pk:
    8
Have the convenience of a fine tip and an ultra fine tip all in one marker. Brightly colored 8-piece set is ideal for research, industrial, biomedical and educational laboratories. When used on glass, metal or porcelain surfaces, ink will not smear when subjected to water. Wash off ink with solvents for repeated use.
  • Specially-formulated, water-resistant ink
  • Assorted colours: Berry; Black; Lime; Navy;Orange; Purple; Sky Blue; Turquoise
  • Fine point tip is compatible with the eCount™