Tisk…

(4-Chlorobenzyl)triphenylphosphonium chloride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL424587-5GEA 946 CZK
SIAL424587-5G
(4-Chlorobenzyl)triphenylphosphonium chloride, Sigma-Aldrich®
(4-Chlorobenzyl)triphenylphosphonium chloride
Vzorec: C₂₅H₂₁Cl₂P
MW: 423,32 g/mol
MDL Number: MFCD00041533
CAS číslo: 1530-39-8
EINECS: 216-228-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR