Tisk…

Digoxin, Supelco®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

D6770-1VLEA 0 CZK
D6770-1VL
Digoxin, Supelco®
Digoxin
Vzorec: C₄₁H₆₄O₁₄
MW: 780,95 g/mol
MDL Number: MFCD00003674
CAS číslo: 20830-75-5
EINECS: 244-068-1
UN: 2811
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR