Iminodiacetic acid
Dodavatel: EHRENSTORFER GMBH
EHERC14285000EA
1640
CZK
EHERC14285000
Iminodiacetic acid
Iminodiacetic acid
Vzorec:
C₄H₇NO₄ MW: 133,10 g/mol |
MDL Number:
MFCD00004280 CAS číslo: 142-73-4 EINECS: 205-555-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...