Tisk…

2-(3-Bromopropoxy)tetrahydropyran, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL415979-5GEA 5270 CZK
SIAL415979-5G
2-(3-Bromopropoxy)tetrahydropyran, Sigma-Aldrich®
2-(3-Bromopropoxy)tetrahydropyran
Vzorec: C₈H₁₅BrO₂
MW: 223,1075 g/mol
MDL Number: MFCD00058593
CAS číslo: 33821-94-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR