Tisk…
Dialysis reservoirs, Spectra/Por®
Dodavatel: Spectrum
SPEC132002EA 2830 CZK
SPEC132002 SPEC132005
Dialysis reservoirs, Spectra/Por®
These cylindrical reservoirs made of polystyrene provide the optimum environment for laboratory dialysis.
The stir-bar creates a vortex that draws the dialysis tubing or ready-to-use dialysis device into the center, maximising dialysis efficiency.