Tisk…

Dimethyl glyoxime disodium salt octahydrate analytical reagent, Supelco®


M-Clarity™ quality level = MQ100

1.03061.0100EA 7530 CZK
1.03061.0100
Dimethyl glyoxime disodium salt octahydrate analytical reagent, Supelco®
Dimethyl glyoxime disodium salt octahydrate
Vzorec: C₄H₆N₂Na₂O₂·8H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150288
CAS číslo: 75006-64-3
EINECS: 262-523-2

Objednat

Výsledky specifikačního testu

Solubility (20 °C) soluble
Molar Mass 304.21 g/mol
Bulk Density 300 kg/m3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR